Assay Details
| Gene Symbol: ZNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013998900.2 | XP_013854354.1 | 2596 | 98 | ||
| XM_021097360.1 | XP_020953019.1 | 2623 | 98 | |||
| XM_021097361.1 | XP_020953020.1 | 2591 | 98 | |||
| XM_021097362.1 | XP_020953021.1 | 2422 | 98 | |||
| XM_021097363.1 | XP_020953022.1 | 2403 | 98 | |||
| XM_021097364.1 | XP_020953023.1 | 2362 | 98 | |||
| XM_021097366.1 | XP_020953025.1 | 2459 | 98 | |||
| XM_021097367.1 | XP_020953026.1 | 2247 | 98 | |||
| XM_021097368.1 | XP_020953027.1 | 2206 | 98 | |||
| XM_021097369.1 | XP_020953028.1 | 2327 | 98 | |||
| GenBank mRNA | AK345950.1 | - | 1 | 1715 | 98 | |
More Information
Set Membership: |
|

ZNF135
Genomic Map