Assay Details
| Gene Symbol: MOXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926931.3 | XP_001926966.2 | 1132 | 65 | ||
| GenBank mRNA | AK346136.1 | - | 7 - 8 | 1173 | 65 | |
| AK349961.1 | - | 7 - 8 | 1207 | 65 | ||
| AK350193.1 | - | 6 - 7 | 839 | 65 | ||
More Information
Set Membership: |
|

MOXD1
Genomic Map