Assay Details
| Gene Symbol: CHST14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121594.4 | XP_003121642.1 | 1929 | 90 | ||
| GenBank mRNA | AK346160.1 | - | 1 | 1924 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CHST14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121594.4 | XP_003121642.1 | 1929 | 90 | ||
| GenBank mRNA | AK346160.1 | - | 1 | 1924 | 90 | |
Set Membership: |
|