Assay Details
| Gene Symbol: AHSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928890.6 | XP_001928925.1 | 719 | 63 | ||
| GenBank mRNA | AK346174.1 | - | 5 - 6 | 693 | 63 | |
| AK347869.1 | - | 5 - 6 | 686 | 63 | ||
| AK391613.1 | - | 5 - 6 | 725 | 63 | ||
| AY609691.1 | - | 5 - 6 | 693 | 63 | ||
More Information
Set Membership: |
|

AHSA1
Genomic Map