Assay Details
| Gene Symbol: LOC110255953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066526.1 | XP_020922185.1 | 76 | |||
| GenBank mRNA | AK346350.1 | - | 3 - 4 | 458 | 76 | |
More Information
Set Membership: |
|

LOC110255953
Genomic Map