Assay Details
| Gene Symbol: LOC110255953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066526.1 | XP_020922185.1 | 651 | 102 | ||
| XM_021066527.1 | XP_020922186.1 | 1005 | 102 | |||
| GenBank mRNA | AK346350.1 | - | 4 - 5 | 635 | 102 | |
More Information
Set Membership: |
|

LOC110255953
Genomic Map