Assay Details
| Gene Symbol: LOC110255953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066526.1 | XP_020922185.1 | 793 | 100 | ||
| XM_021066527.1 | XP_020922186.1 | 1147 | 100 | |||
| GenBank mRNA | AK346350.1 | - | 5 - 6 | 777 | 100 | |
More Information
Set Membership: |
|

LOC110255953
Genomic Map