Assay Details
| Gene Symbol: AK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095842.1 | XP_020951501.1 | 422 | 58 | ||
| XM_021095843.1 | XP_020951502.1 | 319 | 58 | |||
| XM_021095844.1 | XP_020951503.1 | 300 | 58 | |||
| XM_021095846.1 | XP_020951505.1 | 307 | 58 | |||
| GenBank mRNA | AK346409.1 | - | 2 - 3 | 243 | 58 | |
| AK393968.1 | - | 2 - 3 | 261 | 58 | ||
| AY610405.1 | - | 2 - 3 | 235 | 58 | ||
More Information
Set Membership: |
|

AK2
Genomic Map