Assay Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121220.6 | XP_003121268.1 | 450 | 71 | ||
| XM_005659213.2 | XP_005659270.1 | 450 | 71 | |||
| XM_021088683.1 | XP_020944342.1 | 450 | 71 | |||
| GenBank mRNA | AK346435.1 | - | 4 - 5 | 405 | 71 | |
| AK393158.1 | - | 5 - 6 | 598 | 71 | ||
More Information
Set Membership: |
|

HDDC2
Genomic Map