Assay Details
| Gene Symbol: NRBF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925540.5 | XP_001925575.3 | 371 | 63 | ||
| XM_005671025.3 | XP_005671082.2 | 278 | 63 | |||
| XM_005671026.3 | XP_005671083.2 | 390 | 63 | |||
| XM_013983288.2 | XP_013838742.2 | 513 | 63 | |||
| GenBank mRNA | AK346436.1 | - | 3 - 4 | 295 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005671026.3 transcripts.
Set Membership: |
|

NRBF2
Genomic Map