Assay Details
| Gene Symbol: RNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073434.1 | XP_020929093.1 | 371 | 96 | ||
| GenBank mRNA | AK346471.1 | - | 2 - 3 | 340 | 96 | |
| AK390595.1 | - | 2 - 3 | 334 | 96 | ||
More Information
Set Membership: |
|

RNF34
Genomic Map