Assay Details
| Gene Symbol: HAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481441.4 | XP_003481489.1 | 863 | 102 | ||
| XM_021089792.1 | XP_020945451.1 | 1719 | 102 | |||
| XM_021089793.1 | XP_020945452.1 | 895 | 102 | |||
| GenBank mRNA | AK346653.1 | - | 4 - 5 | 1125 | 102 | |
| AY609963.1 | - | 6 - 7 | 814 | 102 | ||
More Information
Set Membership: |
|

HAX1
Genomic Map