Assay Details
| Gene Symbol: CACTIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084097.1 | XP_020939756.1 | 819 | 73 | ||
| XM_021084098.1 | XP_020939757.1 | 819 | 73 | |||
| GenBank mRNA | AK347082.1 | - | 3 - 4 | 787 | 73 | |
More Information
Set Membership: |
|

CACTIN
Genomic Map