Assay Details
| Gene Symbol: TTC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127973.4 | XP_003128021.2 | 468 | 63 | ||
| GenBank mRNA | AK347099.1 | - | 3 - 4 | 437 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TTC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127973.4 | XP_003128021.2 | 468 | 63 | ||
| GenBank mRNA | AK347099.1 | - | 3 - 4 | 437 | 63 | |
Set Membership: |
|