Assay Details
| Gene Symbol: KLHL41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359548.5 | XP_003359596.1 | 1951 | 65 | ||
| GenBank mRNA | AK347246.1 | - | 5 - 6 | 1724 | 65 | |
| AK400874.1 | - | 5 - 6 | 1786 | 65 | ||
More Information
Set Membership: |
|

KLHL41
Genomic Map