Assay Details
| Gene Symbol: SPOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005668871.3 | XP_005668928.2 | 1134 | 69 | ||
| XM_021067137.1 | XP_020922796.1 | 1087 | 69 | |||
| XM_021067138.1 | XP_020922797.1 | 1252 | 69 | |||
| XM_021067139.1 | XP_020922798.1 | 1206 | 69 | |||
| GenBank mRNA | AK347357.1 | - | 8 - 9 | 1068 | 69 | |
| AK390334.1 | - | 9 - 10 | 1134 | 69 | ||
More Information
Set Membership: |
|

SPOP
Genomic Map