Assay Details
| Gene Symbol: CYLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094183.1 | XP_020949842.1 | 2977 | 72 | ||
| XM_021094184.1 | XP_020949843.1 | 3714 | 72 | |||
| XM_021094185.1 | XP_020949844.1 | 3633 | 72 | |||
| XM_021094186.1 | XP_020949845.1 | 4261 | 72 | |||
| XM_021094187.1 | XP_020949846.1 | 2958 | 72 | |||
| XM_021094189.1 | XP_020949848.1 | 3795 | 72 | |||
| XM_021094190.1 | XP_020949849.1 | 3058 | 72 | |||
| XM_021094191.1 | XP_020949850.1 | 3694 | 72 | |||
| XM_021094192.1 | XP_020949851.1 | 5109 | 72 | |||
| XM_021094193.1 | XP_020949852.1 | 2957 | 72 | |||
| GenBank mRNA | AK347366.1 | - | 15 - 16 | 2197 | 72 | |
More Information
Set Membership: |
|

CYLD
Genomic Map