Assay Details
Gene Symbol: EFCAB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013999426.2 | XP_013854880.1 | 1200 | 74 | ||
XM_013999427.2 | XP_013854881.1 | 1200 | 74 | |||
XM_013999428.2 | XP_013854882.1 | 1205 | 74 | |||
XM_013999429.2 | XP_013854883.1 | 1198 | 74 | |||
XM_021096709.1 | XP_020952368.1 | 1201 | 74 | |||
XM_021096710.1 | XP_020952369.1 | 1201 | 74 | |||
XM_021096711.1 | XP_020952370.1 | 1199 | 74 | |||
XM_021096712.1 | XP_020952371.1 | 1199 | 74 | |||
GenBank mRNA | AK347452.1 | - | 1 - 2 | 1183 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
