Assay Details
| Gene Symbol: CCNI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129097.6 | XP_003129145.1 | 1308 | 67 | ||
| XM_021101478.1 | XP_020957137.1 | 898 | 67 | |||
| GenBank mRNA | AK347538.1 | - | 5 - 6 | 894 | 67 | |
| AK391808.1 | - | 5 - 6 | 1031 | 67 | ||
| AY609495.1 | - | 4 - 5 | 503 | 67 | ||
More Information
Set Membership: |
|

CCNI
Genomic Map