Assay Details
| Gene Symbol: NSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013984071.2 | XP_013839525.2 | 9155 | 88 | ||
| XM_021075907.1 | XP_020931566.1 | 9344 | 88 | |||
| XM_021075908.1 | XP_020931567.1 | 9553 | 88 | |||
| XM_021075909.1 | XP_020931568.1 | 8958 | 88 | |||
| XM_021075910.1 | XP_020931569.1 | 9311 | 88 | |||
| XM_021075911.1 | XP_020931570.1 | 9320 | 88 | |||
| XM_021075912.1 | XP_020931571.1 | 9272 | 88 | |||
| XM_021075913.1 | XP_020931572.1 | 9265 | 88 | |||
| XM_021075914.1 | XP_020931573.1 | 8715 | 88 | |||
| XM_021075915.1 | XP_020931574.1 | 9151 | 88 | |||
| XM_021075916.1 | XP_020931575.1 | 9118 | 88 | |||
| GenBank mRNA | AK347594.1 | - | 1 | 425 | 88 | |
More Information
Set Membership: |
|

NSD3
Genomic Map