Assay Details
| Gene Symbol: PARP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126559.5 | XP_003126607.3 | 1143 | 64 | ||
| XM_005664071.3 | XP_005664128.1 | 1345 | 64 | |||
| XM_013997828.2 | XP_013853282.1 | 879 | 64 | |||
| GenBank mRNA | AK347698.1 | - | 7 - 8 | 965 | 64 | |
More Information
Set Membership: |
|

PARP11
Genomic Map