Assay Details
| Gene Symbol: EGFL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005673431.3 | XP_005673488.2 | 492 | 65 | ||
| GenBank mRNA | AK347924.1 | - | 3 - 4 | 742 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: EGFL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005673431.3 | XP_005673488.2 | 492 | 65 | ||
| GenBank mRNA | AK347924.1 | - | 3 - 4 | 742 | 65 | |
Set Membership: |
|