Assay Details
| Gene Symbol: OGFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021078228.1 | XP_020933887.1 | 632 | 61 | ||
| GenBank mRNA | AK348049.1 | - | 6 - 7 | 648 | 61 | |
| AK349490.1 | - | 6 - 7 | 681 | 61 | ||
More Information
Set Membership: |
|

OGFR
Genomic Map