Assay Details
| Gene Symbol: PWWP3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079725.1 | XP_020935384.1 | 137 | 117 | ||
| GenBank mRNA | AK348169.1 | - | 1 - 2 | 99 | 117 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021079725.1 transcripts.
Set Membership: |
|

PWWP3B
Genomic Map