Assay Details
| Gene Symbol: ADAMDEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132811.3 | XP_003132859.2 | 638 | 67 | ||
| XM_021074375.1 | XP_020930034.1 | 639 | 67 | |||
| GenBank mRNA | AK348204.1 | - | 5 - 6 | 495 | 67 | |
More Information
Set Membership: |
|

ADAMDEC1
Genomic Map