Assay Details
| Gene Symbol: EXTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021074088.1 | XP_020929747.1 | 6121 | 92 | ||
| XM_021074089.1 | XP_020929748.1 | 6020 | 92 | |||
| XM_021074090.1 | XP_020929749.1 | 6031 | 92 | |||
| GenBank mRNA | AK348295.1 | - | 1 | 1419 | 92 | |
More Information
Set Membership: |
|

EXTL3
Genomic Map