Assay Details
| Gene Symbol: MRPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005662022.3 | XP_005662079.1 | 155 | 111 | ||
| GenBank mRNA | AK348339.1 | - | 1 - 2 | 99 | 111 | |
| AK392028.1 | - | 1 - 2 | 66 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005662022.3 transcripts.
Set Membership: |
|

MRPS17
Genomic Map