Assay Details
| Gene Symbol: STAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064813.1 | XP_020920472.1 | 260 | 70 | ||
| GenBank mRNA | AK348463.1 | - | 1 - 2 | 237 | 70 | |
| AK390081.1 | - | 1 - 2 | 265 | 70 | ||
More Information
Set Membership: |
|

STAM
Genomic Map