Assay Details
| Gene Symbol: MPHOSPH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097043.1 | XP_020952702.1 | 137 | 88 | ||
| GenBank mRNA | AK348493.1 | - | 1 - 2 | 95 | 88 | |
More Information
Set Membership: |
|

MPHOSPH6
Genomic Map