Assay Details
| Gene Symbol: HTATIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122920.6 | XP_003122968.6 | 521 | 91 | ||
| XM_021083253.1 | XP_020938912.1 | 388 | 91 | |||
| GenBank mRNA | AK348502.1 | - | 3 - 4 | 514 | 91 | |
| AK400232.1 | - | 3 - 4 | 517 | 91 | ||
More Information
Set Membership: |
|

HTATIP2
Genomic Map