Assay Details
| Gene Symbol: DEAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122389.4 | XP_003122437.2 | 1173 | 51 | ||
| XM_021082564.1 | XP_020938223.1 | 1176 | 51 | |||
| GenBank mRNA | AK348534.1 | - | 3 - 4 | 862 | 51 | |
More Information
Set Membership: |
|

DEAF1
Genomic Map