Assay Details
Gene Symbol: INKA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021090027.1 | XP_020945686.1 | 225 | 85 | ||
GenBank mRNA | AK348539.1 | - | 1 - 2 | 229 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021090027.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
