Assay Details
| Gene Symbol: NDUFAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013996214.2 | XP_013851668.2 | 1006 | 66 | ||
| XM_021087630.1 | XP_020943289.1 | 1006 | 66 | |||
| GenBank mRNA | AK348757.1 | - | 8 - 9 | 980 | 66 | |
More Information
Set Membership: |
|

NDUFAF7
Genomic Map