Assay Details
| Gene Symbol: WWTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013992351.2 | XP_013847805.2 | 750 | 57 | ||
| GenBank mRNA | AK348852.1 | - | 3 - 4 | 853 | 57 | |
| AK352300.1 | - | 3 - 4 | 854 | 57 | ||
More Information
Set Membership: |
|

WWTR1
Genomic Map