Assay Details
| Gene Symbol: YOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005667547.3 | XP_005667604.1 | 538 | 94 | ||
| XM_013979638.2 | XP_013835092.1 | 1462 | 94 | |||
| XM_021063331.1 | XP_020918990.1 | 1174 | 94 | |||
| XM_021063333.1 | XP_020918992.1 | 1076 | 94 | |||
| GenBank mRNA | AK349057.1 | - | 1 - 2 | 511 | 94 | |
More Information
Set Membership: |
|

YOD1
Genomic Map