Assay Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076826.1 | XP_020932485.1 | 1727 | 77 | ||
| GenBank mRNA | AK349080.1 | - | 2 - 3 | 776 | 77 | |
More Information
Set Membership: |
|

PLEKHG4B
Genomic Map