Assay Details
| Gene Symbol: SUPT16H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929180.6 | XP_001929215.3 | 3459 | 70 | ||
| XM_021099303.1 | XP_020954962.1 | 3212 | 70 | |||
| GenBank mRNA | AK349089.1 | - | 6 - 7 | 609 | 70 | |
More Information
Set Membership: |
|

SUPT16H
Genomic Map