Assay Details
| Gene Symbol: LYPD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133259.5 | XP_003133307.1 | 3118 | 89 | ||
| XM_013983918.2 | XP_013839372.1 | 3030 | 89 | |||
| XM_013983919.2 | XP_013839373.1 | 3054 | 89 | |||
| XM_021074961.1 | XP_020930620.1 | 3043 | 89 | |||
| XM_021074962.1 | XP_020930621.1 | 3110 | 89 | |||
| GenBank mRNA | AK349094.1 | - | 1 | 916 | 89 | |
More Information
Set Membership: |
|

LYPD6
Genomic Map