Assay Details
| Gene Symbol: SYT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927016.7 | XP_001927051.3 | 3692 | 68 | ||
| GenBank mRNA | AK349145.1 | - | 2 - 3 | 827 | 68 | |
More Information
Set Membership: |
|

SYT10
Genomic Map