Assay Details
| Gene Symbol: TRIM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122865.4 | XP_003122913.1 | 1132 | 89 | ||
| XM_005660994.2 | XP_005661051.1 | 1132 | 89 | |||
| GenBank mRNA | AK349149.1 | - | 2 - 3 | 1110 | 89 | |
| AK389952.1 | - | 2 - 3 | 1103 | 89 | ||
| AK389978.1 | - | 2 - 3 | 89 | |||
| AK393175.1 | - | 2 - 3 | 1116 | 89 | ||
| AK395796.1 | - | 2 - 3 | 1100 | 89 | ||
More Information
Set Membership: |
|

TRIM44
Genomic Map