Assay Details
| Gene Symbol: BST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356861.5 | XP_003356909.2 | 1244 | 78 | ||
| GenBank mRNA | AK349170.1 | - | 6 - 7 | 806 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: BST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356861.5 | XP_003356909.2 | 1244 | 78 | ||
| GenBank mRNA | AK349170.1 | - | 6 - 7 | 806 | 78 | |
Set Membership: |
|