Assay Details
| Gene Symbol: MBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122994.4 | XP_003123042.2 | 393 | 65 | ||
| XM_003353980.3 | XP_003354028.2 | 459 | 65 | |||
| XM_013994842.2 | XP_013850296.2 | 452 | 65 | |||
| XM_013994843.2 | XP_013850297.2 | 379 | 65 | |||
| GenBank mRNA | AK349191.1 | - | 2 - 3 | 279 | 65 | |
| AK352424.1 | - | 2 - 3 | 279 | 65 | ||
More Information
Set Membership: |
|

MBD3
Genomic Map