Assay Details
| Gene Symbol: RPRML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005668698.3 | XP_005668755.1 | 1202 | 95 | ||
| GenBank mRNA | AK349216.1 | - | 1 | 324 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005668698.3 transcripts.
Set Membership: |
|

RPRML
Genomic Map