Assay Details
| Gene Symbol: ZHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088973.1 | XP_020944632.1 | 1150 | 104 | ||
| XM_021088974.1 | XP_020944633.1 | 1150 | 104 | |||
| GenBank mRNA | AK349306.1 | - | 1 - 2 | 68 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021088973.1, XM_021088974.1 transcripts.
Set Membership: |
|

ZHX2
Genomic Map