Assay Details
| Gene Symbol: ST6GALNAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131177.5 | XP_003131225.2 | 426 | 69 | ||
| XM_013989864.2 | XP_013845318.1 | 426 | 69 | |||
| XM_013989867.2 | XP_013845321.1 | 377 | 69 | |||
| GenBank mRNA | AK349463.1 | - | 1 - 2 | 432 | 69 | |
More Information
Set Membership: |
|

ST6GALNAC2
Genomic Map