Assay Details
| Gene Symbol: NAT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095038.1 | XP_020950697.1 | 192 | 67 | ||
| XM_021095039.1 | XP_020950698.1 | 230 | 67 | |||
| GenBank mRNA | AK349518.1 | - | 2 - 3 | 146 | 67 | |
| AK351309.1 | - | 1 - 2 | 279 | 67 | ||
| AK392407.1 | - | 2 - 3 | 168 | 67 | ||
More Information
Set Membership: |
|

NAT14
Genomic Map