Assay Details
Gene Symbol: TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021069307.1 | XP_020924966.1 | 1723 | 71 | ||
XM_021069308.1 | XP_020924967.1 | 3438 | 71 | |||
XM_021069309.1 | XP_020924968.1 | 3312 | 71 | |||
XM_021069310.1 | XP_020924969.1 | 1723 | 71 | |||
XM_021069311.1 | XP_020924970.1 | 1467 | 71 | |||
GenBank mRNA | AK349623.1 | - | 1 - 2 | 176 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
