Assay Details
| Gene Symbol: KRT33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358045.3 | XP_003358093.2 | 1160 | 109 | ||
| GenBank mRNA | AK349769.1 | - | 6 - 7 | 1156 | 109 | |
| AK397182.1 | - | 6 - 7 | 1156 | 109 | ||
| AK397390.1 | - | 6 - 7 | 1156 | 109 | ||
More Information
Set Membership: |
|

KRT33A
Genomic Map