Assay Details
| Gene Symbol: SLC9A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021080387.1 | XP_020936046.1 | 4754 | 99 | ||
| XM_021080388.1 | XP_020936047.1 | 4693 | 99 | |||
| GenBank mRNA | AK349797.1 | - | 1 | 1619 | 99 | |
More Information
Set Membership: |
|

SLC9A7
Genomic Map