Assay Details
| Gene Symbol: TNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128825.4 | XP_003128873.2 | 1034 | 77 | ||
| GenBank mRNA | AK349800.1 | - | 4 - 5 | 982 | 77 | |
More Information
Set Membership: |
|

TNIP2
Genomic Map